Assay Details
| Gene Symbol: Nip7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164472.1 | NP_001157944.1 | 4 | 2755 | 120 | |
| NM_025391.2 | NP_079667.2 | 5 | 2896 | 120 | ||
| NR_028367.1 | 4 | 2989 | 120 | |||
| GenBank mRNA | AK018210.1 | - | 1 | 1049 | 120 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | component of oligomeric golgi complex 8 | Cog8 |
BB235941, C87832 |
-3.0kb | Anti-sense | Chr. 8 - 107048709 - 107056737 on NCBI Build GRCm38 |
| Downstream | transmembrane emp24 protein transport domain containing 6 | Tmed6 |
1810015P03Rik, 1810018I24Rik |
558.0 | Anti-sense | Chr. 8 - 107061484 - 107065644 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nip7
Genomic Map