Assay Details
| Gene Symbol: 6330525I24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018220.1 | - | 1 | 1153 | 61 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 521 | Zfp521 |
B930086A16Rik, Evi3, Znf521 |
-25.2kb | Anti-sense | Chr. 18 - 13687013 - 13972787 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 8430422H06 gene | 8430422H06Rik | 232.8kb | Sense | Chr. 18 - 14232306 - 14278542 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112441 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14018134 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509893 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509857 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509825 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509914 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14018124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509865 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14000812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13917853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13933333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509863 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509843 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13910230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14018123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14018137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14048798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509833 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509842 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509823 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13779768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509844 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509860 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31509859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13933332 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1430411_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430411_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430411_at | - | - |
| Mouse Expression Set 430 | 1430411_at | - | - |
| Mouse Expression Set 430 | 1430411_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430411_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430411_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 757245 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101660035 | - | - |

6330525I24Rik
Genomic Map