Assay Details
| Gene Symbol: 6330571C24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018247.1 | - | 1 | 653 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | EF-hand calcium binding domain 5 | Efcab5 | 4930563A03Rik | -12.0kb | Sense | Chr. 11 - 77089904 - 77199943 on NCBI Build GRCm38 |
| Downstream | slingshot homolog 2 (Drosophila) | Ssh2 |
SSH-2, SSH-2L, mSSH-2L |
3.0kb | Anti-sense | Chr. 11 - 77216169 - 77460220 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6330571C24Rik
Genomic Map