Assay Details
| Gene Symbol: 6330582A15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018259.1 | - | 1 | 1052 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 3010003L21 gene | 3010003L21Rik | -67.2kb | Anti-sense | Chr. 6 - 148946398 - 148947129 on NCBI Build GRCm38 | |
| Downstream | methyltransferase like 20 | Mettl20 |
4833442J19Rik, AI256744 |
125.7kb | Anti-sense | Chr. 6 - 149141513 - 149151171 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6330582A15Rik
Genomic Map