Assay Details
| Gene Symbol: 6430710M23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018303.1 | - | 1 | 354 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DDHD domain containing 2 | Ddhd2 |
2010305K11Rik, SAMWD1, mKIAA0725 |
Sense | Chr. 8 - 25725324 - 25754590 on NCBI Build GRCm38 | |
| Downstream | BCL2-associated athanogene 4 | Bag4 |
2410112I15Rik, SODD |
19.0kb | Sense | Chr. 8 - 25764538 - 25785209 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6430710M23Rik
Genomic Map