Assay Details
| Gene Symbol: 6530403M18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018320.1 | - | 1 | 552 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | small integral membrane protein 10 like 2A | Smim10l2a |
6330419J24Rik, Ncrna00086 |
-2.7kb | Sense | Chr. X - 56374586 - 56378470 on NCBI Build GRCm38 |
| Downstream | DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 26B | Ddx26b |
4930535D10Rik, 6330505F04Rik, D130066O12 |
72.2kb | Sense | Chr. X - 56454821 - 56507843 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6530403M18Rik
Genomic Map