Assay Details
| Gene Symbol: 9030409K20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018498.1 | - | 1 | 1452 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | myotrophin | Mtpn |
5033418D15Rik, Gcdp, V1 |
-343.4kb | Sense | Chr. 6 - 35508824 - 35539888 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 9330158H04 gene | 9330158H04Rik | 447.7kb | Sense | Chr. 6 - 36333138 - 36388234 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9030409K20Rik
Genomic Map