Assay Details
| Gene Symbol: 9030420N05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018521.1 | - | 1 | 1751 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Tctex1 domain containing 2 | Tctex1d2 |
0610012D17Rik, 2410011K15Rik |
-7.6kb | Sense | Chr. 16 - 32419686 - 32429099 on NCBI Build GRCm38 |
| Downstream |
solute carrier family 51, alpha subunit |
Slc51a |
AV001382, AW261577, D630035O19Rik, OSTalpha, Osta |
36.6kb | Anti-sense | Chr. 16 - 32475578 - 32487879 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9030420N05Rik
Genomic Map