Assay Details
| Gene Symbol: 9030601B04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018532.1 | - | 1 | 949 | 85 | |
| BG072497.2 | - | 1 | 85 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ATPase, H+ transporting, lysosomal V0 subunit C, pseudogene 2 |
Atp6v0c-ps2 |
Atp6l-ps2, Gm15487 |
-133.9kb | Sense | Chr. 6 - 37805975 - 37807107 on NCBI Build GRCm38 |
| Downstream | SV2 related protein homolog (rat)-like | Svopl | 9430071P14Rik | 41.3kb | Anti-sense | Chr. 6 - 37983739 - 38047014 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9030601B04Rik
Genomic Map