Assay Details
| Gene Symbol: 9030622M22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018564.1 | - | 1 | 948 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chloride channel 3 | Clcn3 | Clc3 | Sense | Chr. 8 - 60910389 - 60983311 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA B230317F23 gene | B230317F23Rik | 18.3kb | Anti-sense | Chr. 8 - 60983371 - 60987512 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9030622M22Rik
Genomic Map