Assay Details
| Gene Symbol: Zbtb7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018596.1 | - | 1 | 657 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitogen-activated protein kinase kinase 2 | Map2k2 |
AA589381, MEK2, MK2, Prkmk2 |
-26.6kb | Sense | Chr. 10 - 81105912 - 81124697 on NCBI Build GRCm38 |
| Downstream | protein inhibitor of activated STAT 4 | Pias4 |
PIASY, Pias-gamma, Piasg |
1.7kb | Anti-sense | Chr. 10 - 81153961 - 81167720 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zbtb7a
Genomic Map