Assay Details
| Gene Symbol: Airn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK051797.1 | - | 1 | 42 | 110 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 22 (organic cation transporter), member 1 |
Slc22a1 |
Lx1, Oct1, Orct, Orct1 |
-87.5kb | Anti-sense | Chr. 17 - 12648874 - 12675838 on NCBI Build GRCm38 |
| Downstream | MAS1 oncogene | Mas1 |
Mas-1, Mgra |
74.8kb | Anti-sense | Chr. 17 - 12838841 - 12868145 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Airn
Genomic Map