Assay Details
| Gene Symbol: Gm12273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052659.1 | - | 1 - 2 | 159 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | A kinase (PRKA) anchor protein 10 | Akap10 |
1500031L16Rik, B130049N18Rik, D-AKAP-2, D-AKAP2, PRKA10 |
-20.6kb | Anti-sense | Chr. 11 - 61871307 - 61930307 on NCBI Build GRCm38 |
| Downstream | sperm antigen with calponin homology and coiled-coil domains 1 | Specc1 |
2810012G08Rik, B230396K10Rik, Cytsb |
5.1kb | Sense | Chr. 11 - 61956763 - 62223013 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm12273
Genomic Map