Assay Details
| Gene Symbol: D630022N01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052681.1 | - | 1 - 2 | 1273 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | guanine monophosphate synthetase | Gmps |
AA591640, AI047208 |
Sense | Chr. 3 - 63964819 - 64021490 on NCBI Build GRCm38 | |
| Downstream |
vomeronasal 2, receptor 1 |
Vmn2r1 |
Casrl1, EG56544, V2r83 |
111.1kb | Sense | Chr. 3 - 64079774 - 64220847 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

D630022N01Rik
Genomic Map