Assay Details
| Gene Symbol: Snapc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK053153.1 | - | 1 | 543 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 11413 | Gm11413 | OTTMUSG00000000829 | -72.6kb | Anti-sense | Chr. 4 - 83378317 - 83390668 on NCBI Build GRCm38 |
| Downstream | coiled-coil domain containing 171 | Ccdc171 |
4930418J05Rik, 4930473A06Rik, A330015D16Rik |
59.7kb | Sense | Chr. 4 - 83525537 - 83866143 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snapc3
Genomic Map