Assay Details
Gene Symbol: 9530036O11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK053224.1 | - | 626 | 101 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Downstream | ring finger protein 32 | Rnf32 |
1700009J01Rik, 2700025B22Rik, 4930542N22Rik, Lmbr2 |
261.2kb | Sense | Chr. 5 - 29195170 - 29225526 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
