Assay Details
| Gene Symbol: 4930519P11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015848.1 | - | 1 | 1392 | 99 | |
| AK053350.1 | - | 1 | 1347 | 99 | ||
| AV259580.2 | - | 1 | 388 | 99 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | peroxisomal membrane protein 4 | Pxmp4 |
3010018P03Rik, Pmp24 |
-8.2kb | Sense | Chr. 2 - 154587044 - 154603673 on NCBI Build GRCm38 |
| Downstream | charged multivesicular body protein 4B | Chmp4b |
2010012F05Rik, C76846, Snf7-2 |
43.6kb | Anti-sense | Chr. 2 - 154657026 - 154694783 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930519P11Rik
Genomic Map