Assay Details
| Gene Symbol: Ipmk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076771.1 | - | 165 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CDGSH iron sulfur domain 1 | Cisd1 |
AU043990, AW743335, D10Ertd214e, Zcd1, mitoNEET |
-36.9kb | Anti-sense | Chr. 10 - 71330494 - 71344849 on NCBI Build GRCm38 |
| Downstream | hematological and neurological expressed 1-like pseudogene | 1700049L16Rik | 577.5kb | Anti-sense | Chr. 10 - 71979885 - 71980690 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Ipmk
Genomic Map