Assay Details
| Gene Symbol: Junos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076859.1 | - | 2 - 3 | 163 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | jun proto-oncogene | Jun |
AP-1, Junc, c-jun |
-94.9kb | Anti-sense | Chr. 4 - 95049036 - 95052222 on NCBI Build GRCm38 |
| Downstream | predicted gene 12708 | Gm12708 | OTTMUSG00000008048 | 45.8kb | Sense | Chr. 4 - 95193274 - 95209724 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Junos
Genomic Map