Assay Details
| Gene Symbol: Tmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081111.2 | NP_001074580.1 | 17 | 4810 | 101 | |
| GenBank mRNA | AK078561.1 | - | 1 | 1146 | 101 | |
| AK136794.1 | - | 1 | 1153 | 101 | ||
| AK161423.1 | - | 1 | 1144 | 101 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | EGF domain-specific O-linked N-acetylglucosamine (GlcNAc) transferase | Eogt |
A130022J15Rik, AI447490, AW214473, AW259391, Aer61 |
-3.5kb | Sense | Chr. 6 - 97110016 - 97149703 on NCBI Build GRCm38 |
| Downstream | ubiquitin-like modifier activating enzyme 3 | Uba3 |
A830034N06Rik, AI256736, AI848246, AW546539, Ube1c |
28.6kb | Sense | Chr. 6 - 97183632 - 97205647 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tmf1
Genomic Map