Assay Details
| Gene Symbol: 6230424C14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK078961.1 | - | 1 | 1053 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | integrin alpha 2 | Itga2 |
CD49B, DX5 |
-21.1kb | Sense | Chr. 13 - 114835912 - 114932041 on NCBI Build GRCm38 |
| Downstream | integrin alpha 1 | Itga1 |
CD49A, E130012M19Rik, Vla1 |
3.3kb | Sense | Chr. 13 - 114958079 - 115101964 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6230424C14Rik
Genomic Map