Assay Details
| Gene Symbol: LOC432885 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK079079.1 | - | 1 | 3347 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protocadherin 9 | Pcdh9 | Sense | Chr. 14 - 93013700 - 93890721 on NCBI Build GRCm38 | ||
| Downstream |
predicted gene, 33298 |
Gm33298 | 206.7kb | Sense | Chr. 14 - 93861481 - 93872573 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC432885
Genomic Map