Assay Details
| Gene Symbol: Pcgf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK079197.1 | - | 1 | 1347 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ankyrin repeat domain 1 (cardiac muscle) | Ankrd1 |
Alrp, CARP, Crap, MARP1 |
-339.1kb | Anti-sense | Chr. 19 - 36111965 - 36119844 on NCBI Build GRCm38 |
| Downstream | HECT domain containing 2 | Hectd2 |
4921524L07, A630025O09Rik, AW212605 |
93.5kb | Sense | Chr. 19 - 36553969 - 36621135 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pcgf5
Genomic Map