Assay Details
| Gene Symbol: Fam188a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK079720.1 | - | 1 | 245 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA E030013I19 gene | E030013I19Rik | -73.6kb | Anti-sense | Chr. 2 - 12301246 - 12312315 on NCBI Build GRCm38 | |
| Downstream | phosphotriesterase related | Pter |
AI790318, Mpr56-1 |
537.3kb | Anti-sense | Chr. 2 - 12924034 - 13003453 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fam188a
Genomic Map