Assay Details
| Gene Symbol: A930037O16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK080826.1 | - | 1 | 1750 | 100 | |
| BB311547.2 | - | 1 | 359 | 100 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | G protein-coupled receptor 37 | Gpr37 |
AI848630, Pael-R |
-48.5kb | Sense | Chr. 6 - 25668523 - 25689980 on NCBI Build GRCm38 |
| Downstream | protection of telomeres 1A | Pot1a |
1500031H18Rik, AI851169, Pot1 |
3.2kb | Sense | Chr. 6 - 25743735 - 25809277 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A930037O16Rik
Genomic Map