Assay Details
| Gene Symbol: A230072E10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015600.2 | 1 - 2 | 1392 | 92 | ||
| GenBank mRNA | AK081752.1 | - | 1 - 2 | 1392 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | WNK lysine deficient protein kinase 3 | Wnk3 |
Prkwnk3, Wnk3-ps |
-13.7kb | Anti-sense | Chr. X - 151168777 - 151320194 on NCBI Build GRCm38 |
| Downstream |
family with sequence similarity 120, member C |
Fam120c |
D930001I21Rik, ORF34 |
1.1kb | Anti-sense | Chr. X - 151342932 - 151474136 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

A230072E10Rik
Genomic Map