Assay Details
| Gene Symbol: A230072E10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015600.2 | 2 - 3 | 1487 | 64 | ||
| GenBank mRNA | AK081752.1 | - | 2 - 3 | 1487 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | WNK lysine deficient protein kinase 3 | Wnk3 |
Prkwnk3, Wnk3-ps |
Anti-sense | Chr. X - 151168777 - 151320194 on NCBI Build GRCm38 | |
| Downstream |
family with sequence similarity 120, member C |
Fam120c |
D930001I21Rik, ORF34 |
8.9kb | Anti-sense | Chr. X - 151342932 - 151474136 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2328171 | AK081752.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2328171 | NR_015600.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P417677 | BC025887.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P417677 | BC137973.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P417677 | BC171940.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P417677 | AY150024.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P417677 | AY150023.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P417677 | BC025887.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P417677 | BC137973.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P417677 | BC171940.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P417677 | AY150024.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P417677 | AY150023.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 110010 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539541 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 418342 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 671505 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 83600 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 841559 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 42077 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668595 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 11347 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 389437 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931051 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 516204 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 960326 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 638267 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730837 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 144588 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 168589 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 119130 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 177061 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136026 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146403 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112846 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118392 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102507 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09459164 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102507 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03776787 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136026 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112846 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146403 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09459105 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118392 | - | - |
| Whole Mouse Genome G4122A | A_51_P461082 | AK038361.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P265643 | AK081752.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P265643 | NR_015600.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P357314 | AK038885.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107196 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146403 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118392 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102507 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136026 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744811 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14811724 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15042284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32745016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32745003 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14896590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14553056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744825 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744899 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32745008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14852477 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744808 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744803 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15124199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32745002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14921062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32745017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744809 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32745005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744823 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744800 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744797 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14878035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744790 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744916 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32745010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15170144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32745004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744984 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32745006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744818 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14885866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744880 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32745013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14821671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15042288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32745007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744827 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32744795 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10607331 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10607330 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00020000.mm.1 | NR_015600.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00020000.mm.1 | TC0X00003178.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 334581 | - | - |

A230072E10Rik
Genomic Map