Assay Details
| Gene Symbol: Pgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK083181.1 | - | 1 | 1947 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 32014 |
Gm32014 | -158.5kb | Sense | Chr. 9 - 8644078 - 8740988 on NCBI Build GRCm38 | |
| Downstream | Rho GTPase activating protein 42 | Arhgap42 |
9030420J04Rik, BE136619 |
92.5kb | Sense | Chr. 9 - 8994331 - 9239164 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pgr
Genomic Map