Assay Details
| Gene Symbol: Iqcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK083217.1 | - | 1 | 652 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | doublecortin domain containing 2b | Dcdc2b |
Gm12964, Gm1366, OTTMUSG00000009523 |
-3.6kb | Sense | Chr. 4 - 129608331 - 129614477 on NCBI Build GRCm38 |
| Downstream | coiled coil domain containing 28B | Ccdc28b |
1110002E08Rik, 1810010N17Rik |
164.0 | Sense | Chr. 4 - 129619274 - 129623908 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Iqcc
Genomic Map