Assay Details
| Gene Symbol: E230001N04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK083527.1 | - | 1 | 52 | 61 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FK506 binding protein 5 | Fkbp5 |
D17Ertd592e, Dit1, FKBP-5, FKBP51 |
-37.1kb | Anti-sense | Chr. 17 - 28398753 - 28486149 on NCBI Build GRCm38 |
| Downstream | armadillo repeat containing 12 | Armc12 |
4930511I11Rik, AV041658 |
3.6kb | Sense | Chr. 17 - 28527534 - 28539110 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

E230001N04Rik
Genomic Map