Assay Details
| Gene Symbol: 2010007H06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK084979.1 | - | 1 | 2842 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 684 | Gm684 | -1.1kb | Anti-sense | Chr. 9 - 51270258 - 51279729 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 1810046K07 gene | 1810046K07Rik | 5.5kb | Anti-sense | Chr. 9 - 51289686 - 51328924 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2010007H06Rik
Genomic Map