Assay Details
| Gene Symbol: Gm5112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK085200.1 | - | 1653 | 113 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 2 (facilitated glucose transporter), member 3 |
Slc2a3 |
AA408729, AL023014, AL024341, AU040424, C78366, Glut-3, Glut3 |
-10.4kb | Anti-sense | Chr. 6 - 122727809 - 122802270 on NCBI Build GRCm38 |
| Downstream | forkhead box J2 | Foxj2 | Fhx | 7.1kb | Sense | Chr. 6 - 122820184 - 122845366 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm5112
Genomic Map