Assay Details
| Gene Symbol: Gm8149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK085632.1 | - | 410 | 89 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
wingless-type MMTV integration site family, member 11 |
Wnt11 | -42.7kb | Sense | Chr. 7 - 98834921 - 98854747 on NCBI Build GRCm38 | |
| Downstream | diacylglycerol O-acyltransferase 2 | Dgat2 |
0610010B06Rik, ARAT, DGAT-2 |
254.8kb | Anti-sense | Chr. 7 - 99153663 - 99182713 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm8149
Genomic Map