Assay Details
| Gene Symbol: Csnk1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK086172.1 | - | 1 | 1250 | 96 | |
| BC040394.1 | - | 2 | 2796 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 16 (monocarboxylic acid transporters), member 3 |
Slc16a3 |
Mct3, Mct4 |
-19.4kb | Anti-sense | Chr. 11 - 120948484 - 120959000 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 30062 |
Gm30062 | 10.9kb | Anti-sense | Chr. 11 - 120991438 - 121015421 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Csnk1d
Genomic Map