Assay Details
| Gene Symbol: 5730424H11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK030731.1 | - | 1 | 2139 | 97 | |
| AK086470.1 | - | 1 | 2142 | 97 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6958 | Mir6958 |
Gm27581, mmu-mir-6958 |
-10.6kb | Anti-sense | Chr. 15 - 89185465 - 89185537 on NCBI Build GRCm38 |
| Downstream |
protein phosphatase 6, regulatory subunit 2 |
Ppp6r2 |
1110033O10Rik, 8430411H09Rik, B230107H12Rik, Pp6r2, Saps2 |
13.1kb | Sense | Chr. 15 - 89211521 - 89287015 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5730424H11Rik
Genomic Map