Assay Details
| Gene Symbol: A830019L24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040551.1 | 2 - 3 | 487 | 170 | ||
| GenBank mRNA | AK043679.1 | - | 2 - 3 | 488 | 170 | |
| AK086499.1 | - | 122 | 170 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 9530052C20 gene | 9530052C20Rik | -345.5kb | Sense | Chr. 3 - 142878399 - 142898015 on NCBI Build GRCm38 | |
| Downstream | predicted gene 6260 | Gm6260 | EG621809 | 414.1kb | Anti-sense | Chr. 3 - 143670892 - 143712143 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

A830019L24Rik
Genomic Map