Assay Details
| Gene Symbol: Acnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164565.1 | NP_001158037.1 | 3 | 2425 | 104 | |
| GenBank mRNA | AK050104.1 | - | 1 | 1043 | 104 | |
| AK086578.1 | - | 1 | 1044 | 104 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | acyl-coenzyme A amino acid N-acyltransferase 2 | Acnat2 | C730036D15Rik | -35.8kb | Sense | Chr. 4 - 49375341 - 49408151 on NCBI Build GRCm38 |
| Downstream | bile acid-Coenzyme A: amino acid N-acyltransferase | Baat |
AI118337, AI158864, BAT |
42.2kb | Sense | Chr. 4 - 49489416 - 49507912 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Acnat1
Genomic Map