Assay Details
| Gene Symbol: Sirt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK086910.1 | - | 1 | 2354 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | resistance to inhibitors of cholinesterase 8 homolog (C. elegans) | Ric8 |
AI114950, RIC-8, Ric8a |
-10.1kb | Anti-sense | Chr. 7 - 140857397 - 140863731 on NCBI Build GRCm38 |
| Downstream |
proteasome (prosome, macropain) 26S subunit, non-ATPase, 13 |
Psmd13 | S11 | 6.0kb | Anti-sense | Chr. 7 - 140882394 - 140898642 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sirt3
Genomic Map