Assay Details
| Gene Symbol: Sfta2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163194.1 | NP_001156666.1 | 2 - 3 | 305 | 78 | |
| GenBank mRNA | AK087178.1 | - | 2 - 3 | 306 | 78 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sfta2
Genomic Map