Assay Details
| Gene Symbol: A630035D09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK041756.1 | - | 1 | 2147 | 66 | |
| AK087629.1 | - | 1 | 1444 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | BCL2-like 1 | Bcl2l1 |
Bcl(X)L, Bcl-XL, Bcl2l, BclX, bcl-x, bcl2-L-1 |
Sense | Chr. 2 - 152758732 - 152832581 on NCBI Build GRCm38 | |
| Downstream |
TPX2, microtubule-associated protein homolog (Xenopus laevis) |
Tpx2 |
2610005B21Rik, DIL2, REPP86, p100 |
36.8kb | Anti-sense | Chr. 2 - 152847964 - 152895321 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A630035D09Rik
Genomic Map