Assay Details
| Gene Symbol: F830002E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK089566.1 | - | 1 | 1246 | 79 | |
| AK172269.1 | - | 1 | 1246 | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neurensin 1 | Nrsn1 |
Neurensin-1, Neuro-p24, Vmp |
-158.1kb | Sense | Chr. 13 - 25252039 - 25270070 on NCBI Build GRCm38 |
| Downstream | predicted gene 11351 | Gm11351 | OTTMUSG00000000620 | 490.9kb | Sense | Chr. 13 - 25921033 - 25966316 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

F830002E08Rik
Genomic Map