Assay Details
| Gene Symbol: 1700025D23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK131679.1 | - | 1 - 2 | 434 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sidekick homolog 2 (chicken) | Sdk2 |
4632412F08Rik, 5330435L01Rik, Sdk-2, mKIAA1514 |
-25.3kb | Anti-sense | Chr. 11 - 113776374 - 114066181 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700092K14 gene | 1700092K14Rik | 105.6kb | Sense | Chr. 11 - 114198258 - 114199199 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

1700025D23Rik
Genomic Map