Assay Details
| Gene Symbol: Gm15412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046043.1 | 2 - 3 | 822 | 95 | ||
| GenBank mRNA | AK131755.1 | - | 2 - 3 | 823 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6394 | Mir6394 |
Gm27929, mmu-mir-6394 |
-33.7kb | Anti-sense | Chr. 7 - 96305667 - 96305765 on NCBI Build GRCm38 |
| Downstream | predicted gene 15413 | Gm15413 |
ENSMUSG00000053049, OTTMUSG00000021246 |
450.7kb | Sense | Chr. 7 - 96791433 - 96801549 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm15412
Genomic Map