Assay Details
| Gene Symbol: Fhitos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK131787.1 | - | 2 - 3 | 449 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Mnd1 retrotransposed pseudogene | Mnd1-ps | Gm3833 | -872.9kb | Sense | Chr. 14 - 9884095 - 9886868 on NCBI Build GRCm38 |
| Downstream | microRNA 5124b | Mir5124b |
Gm27681, mmu-mir-5124b |
504.9kb | Sense | Chr. 14 - 11282477 - 11282561 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Fhitos
Genomic Map