Assay Details
Gene Symbol: Kcnd3os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040759.1 | 1 - 2 | 402 | 97 | ||
GenBank mRNA | AK131816.1 | - | 1 - 2 | 402 | 97 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 34781 |
Gm34781 | -353.1kb | Anti-sense | Chr. 3 - 105094996 - 105098403 on NCBI Build GRCm38 | |
Downstream | DEAD (Asp-Glu-Ala-Asp) box polypeptide 20 | Ddx20 |
GEMIN3, dp103 |
225.3kb | Sense | Chr. 3 - 105678270 - 105688113 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
