Assay Details
| Gene Symbol: Kcnd3os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040759.1 | 2 - 3 | 564 | 86 | ||
| GenBank mRNA | AK131816.1 | - | 2 - 3 | 564 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 34781 |
Gm34781 | -352.7kb | Anti-sense | Chr. 3 - 105094996 - 105098403 on NCBI Build GRCm38 | |
| Downstream |
potassium voltage-gated channel, Shal-related family, member 3 |
Kcnd3 |
AW045978, Kncd3, Kv4.3 |
423.0 | Anti-sense | Chr. 3 - 105452093 - 105674002 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnd3os
Genomic Map