Assay Details
| Gene Symbol: Npw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099664.1 | NP_001093134.1 | 1 - 2 | 429 | 64 | |
| GenBank mRNA | AK131881.1 | - | 1 - 2 | 211 | 64 | |
More Information
Set Membership: |
|

Npw
Genomic Map