Assay Details
| Gene Symbol: 4930544I03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK132687.1 | - | 437 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 26512 |
Gm26512 | -29.0kb | Anti-sense | Chr. 12 - 90738212 - 90812245 on NCBI Build GRCm38 | |
| Downstream | centrosomal protein 128 | Cep128 |
4930534B04Rik, 5430424K18Rik, AI449708, AI480494, E230024F20 |
142.4kb | Sense | Chr. 12 - 90998490 - 91384416 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930544I03Rik
Genomic Map