Assay Details
| Gene Symbol: Gm11978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028586.1 | 2 - 3 | 238 | 71 | ||
| GenBank mRNA | AK132798.1 | - | 2 - 3 | 239 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | receptor (calcitonin) activity modifying protein 3 | Ramp3 | AI850306 | Anti-sense | Chr. 11 - 6658533 - 6677476 on NCBI Build GRCm38 | |
| Downstream | predicted gene 11981 | Gm11981 | OTTMUSG00000005129 | 56.4kb | Sense | Chr. 11 - 6716001 - 6720037 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm11978
Genomic Map