Assay Details
| Gene Symbol: Gm11978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028586.1 | 5 - 6 | 555 | 92 | ||
| GenBank mRNA | AK132798.1 | - | 5 - 6 | 556 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | whey acidic protein | Wap | -11.5kb | Sense | Chr. 11 - 6635483 - 6638649 on NCBI Build GRCm38 | |
| Downstream | receptor (calcitonin) activity modifying protein 3 | Ramp3 | AI850306 | 7.3kb | Anti-sense | Chr. 11 - 6658533 - 6677476 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm11978
Genomic Map